Precellys® MK28 Lysing Kit, Hard Tissue Grinding, Bertin
Supplier: BERTIN CORP
P000910-LYSK0-A.0 P000917-LYSK0-A.0 P000925-LYSK0-A.0
10144-492EA 406.93 CAD
10144-492 10144-518 10144-558
Precellys® MK28 Lysing Kit, Hard Tissue Grinding, Bertin
Homogenizers Tissue Grinders
MK28 is made with inert 2.8 mm stainless steel beads
  • Ideal for teeth, hair, seeds, bones, etc. grinding
  • Dnase/Rnase free
  • Inert 2.8 mm stainless steel beads
  • Autoclavable
  • Usable with Precellys®24, Precellys® Evolution and Minilys® homogenizers
The custom designed reinforced tubes are intended for bead beating applications, and feature O-Rings and reinforced materials for dry grinding valuable or very hard samples.
It is designed for hard tissue grinding like muscle, skin, spincor, bone, teeth, hair, seeds, corn, rice and more.
Order Now
SPECIFICATIONS
Environmentally Preferable