Minilys Homogenizer, Bertin
Supplier: Bertin
P000673-MLYS0A
10144-650EA 5200 CAD
10144-650
Minilys Homogenizer, Bertin
Homogenizers Benchtop Homogenizers
The Minilys is a compact and efficient homogenizer dedicated to sample preparation
  • 3 speed settings: 3000, 4000 and 5000 rpm
  • Manual tube locking system
  • Capacity: three tubes of 0.5, 1.5 or 2mL and one tube of 7mL
  • Simple user interface with two parameters (speed and time)
The unit allows users to homogenize, grind, or lyse any type of sample in as little as 30 seconds, due to 3D motion movement. The Minilys Homogenizer is ideally suited for daily use in sample homogenization.
Certifications: CE certified. FCC part 15 certified.