Anti-CHKA Rabbit Polyclonal Antibody

Supplier: Abgent
AP7064A
60942-914EA 1265.83 CAD
60942-914
Anti-CHKA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CHKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (50–150 aa)
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR