Anti-PROX1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2035B
80088-356EA 1265.83 CAD
80088-356
Anti-PROX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PROX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal ( 531–546 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR