Anti-NCS1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1551C
80088-128EA 1265.83 CAD
80088-128
Anti-NCS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center ( 157–168 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR