Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: 5A8E5]

Supplier: Genscript
A00187-100
89493-648EA 246.5 CAD
89493-648
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: 5A8E5]
Antibodies
THE™ DYKDDDDK Tag antibody, mAb, Mouse recognizes DYKDDDDK; tags localized at the C-terminal, N-terminal and internal region of DYKDDDDK
tagged fusion proteins.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR