VWR® Spouted Collection Containers
Supplier: VWR
VWR®
15704-047
15704-047CS 1063.92 CAD
15704-047
VWR® Spouted Collection Containers
Sample Containers General Purpose Containers
These 237 mL (8 oz.) containers are manufactured out of polystyrene with excellent clarity.
  • Graduated markings
  • Polystyrene construction
  • Convenient pour spout
  • Superior clarity
Convenient pour spout for simplified specimen retrieval. Each container is graduated in 10 mL (cc) and 0.5 oz. increments up to 6.5 oz.