VWR® Carboys with 3" Sanitary Neck, PP
Supplier: VWR
VWR®
155-2131
89170-788EA 776.57 CAD
89170-788
VWR® Carboys with 3" Sanitary Neck, PP
Bottles Carboys
VWR® polypropylene carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
  • Polypropylene construction
  • Translucent, autoclavable resin molding
The distinctive rectangular shape saves valuable bench space.
VWR® polypropylene carboys are molded from a translucent USP Class VI, FDA grade, autoclavable resin. The resin offers good chemical resistance and heat stability. Polypropylene carboys, as well as all versatile caps, adapters, versatile barbs, and spigots may be autoclaved. The recommended autoclaving cycle is 20 minutes at 15 psi. and 121 °C (250 °F).
Made in the USA.
Large handles on top of the container and molded handles in the base provide a full, secure grip to make lifting, carrying, and pouring easy. Sanitary carboys feature a 7.6 cm (3") sanitary neck. All carboys have their material and volume clearly imprinted on the outside of the bottle, as well as large, easy to read metric graduation marks which are certified to ±5% accuracy.