Hydranal® Coulometric Karl Fischer Titration Reagents, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
34821-6X50ML 34840-6X50ML 34843-6X500ML-US 34868-100ML 34810-6X500ML-US 34836-6X1L-US 34868-500ML-US 34868-6X100ML
BJ34810-500MLCS 1428.53 CAD
BJ34810-500ML BJ34820-500ML BJ34821-50ML BJ34840-50ML BJ34843-500ML BJ34868-500ML BJ34868-100ML BJ34810500MLUS BJ34836-1L-US BJ34868-500MLS BJ34868-6100ML
Hydranal® Coulometric Karl Fischer Titration Reagents, Honeywell Research Chemicals
Karl Fischer Titrants and Reagents
Coulometric Karl Fischer titrations normally require two reagent solutions: an anolyte (the solution in the anodic compartment) and a catholyte (the solution in the cathodic compartment).
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range available
  • Long shelf life (up to five years)
Order Now
SPECIFICATIONS
Environmentally Preferable