PAGE-Optimizer™ Spin Columns, G-Biosciences
Supplier: G-Biosciences
786-947 786-948
CA10781-906EA 143.69 CAD
CA10781-906 CA10781-908
PAGE-Optimizer™ Spin Columns, G-Biosciences
Filters Centrifugal Filters
PAGE-Optimizer is a unique, spin column for the preparation of samples for SDS-PAGE. The columns contain a proprietary separation matrix that removes contaminants such as salts, detergents, cellular agents and other common buffering agents.
  • Just spin to a perfect gel
  • 5 minute protocol
  • Clean up samples for SDS-PAGE for improved band resolution
PAGE-Optimizer helps eliminate smeared lanes, distorted and/ or smiling bands and swollen lanes. PAGE-Optimizer has also been shown to reveal protein bands previously masked by the above interfering agents.