Face Shield, Unimed-Midwest
Supplier: Unimed
SSDS00205B
89068-863CS 2100.12 CAD
89068-863
Face Shield, Unimed-Midwest
Face Shields
Splash shield provides comfortable face protection.
  • Soft elastic strap wraps around head to secure shield
  • Anti-fog
  • Optically Clear
  • 13 inch impenetrable foam barrier for wide coverage
  • Disposable and single use
It is not recommended to disinfect and reuse shields because doing so would destroy the anti-fog coating.