YMC-Pack Pro C18 HPLC Columns, YMC AMERICA, INC
Supplier: YMC America,Inc
AS12S03-0104WFG AS12S03-0501WT AS12S03-0502WT AS12S03-0503WT AS12S03-0506WT AS12S03-0546WT AS12S03-1001WT AS12S03-1002WT AS12S03-1003WT AS12S03-1006WT AS12S03-1010WT AS12S03-1046WT AS12S03-1502WT AS12S03-1503WT AS12S03-1510WT AS12S03-1546WT AS12S03-2503WT AS12S03-2546WT AS12S03-H501WT AS12S03-H502WT AS12S03-H546WT AS12S03-L502WT AS12S03-L506WT AS12S03-L546WT AS12S05-0310WTG AS12S05-0502WT AS12S05-0503WT AS12S05-0520WTG AS12S05-0520WX AS12S05-0530WTG AS12S05-0546WT AS12S05-1002WT AS12S05-1003WT AS12S05-1020WT AS12S05-1020WX AS12S05-1030WT AS12S05-1046WT AS12S05-1501WT AS12S05-1502WT AS12S05-1503WT AS12S05-1506WT AS12S05-1510WT AS12S05-1520WT AS12S05-1530WT AS12S05-1546WT AS12S05-2501WT AS12S05-2546WT
97029-242EA 1284.48 CAD
97029-242 97029-250 97029-254 97029-258 97029-262 97029-266 97029-270 97029-272 97029-276 97029-280 97029-282 97029-284 97029-288 97029-292 97029-296 97029-300 97029-302 97029-304 97029-312 97029-316 97029-328 97029-330 97029-332 97029-334 97029-336 97029-338 97029-342 97029-358 97029-362 97029-366 97029-370 97029-372 97029-374 97029-378 97029-382 97029-386 97029-388 97029-390 97029-392 97029-396 97029-398 97029-402 97029-406 97029-408 97029-410 97029-412 97029-414 97029-418 97029-422 97029-440
YMC-Pack Pro C18 HPLC Columns, YMC AMERICA, INC
Chromatography Columns
These columns use tightly controlled, highly purified silica, a high-coverage C18 bonding, and a unique end-capping procedure utilizing Lewis acid/Lewis base chemistry.
  • Made with highly inert, ultrapure, pH neutral silica
  • Very high efficiency columns
These features provide unequaled separations with sharp and symmetrical peaks for all types of organic molecules, especially basic pharmaceuticals and chelating compounds.