Cryo Hold™ TS818 Cryogenic Thermal Transfer Label Kit
Supplier: Computype
1020386 1020388-1 1020388 1020390-1 1020390 15298937 1020386-1
10769-944KT 1475.26 CAD
10769-944 10769-946 10769-948 10769-950 10769-952 10769-954 10769-956 10770-058
Cryo Hold™ TS818 Cryogenic Thermal Transfer Label Kit
Labels
Cryo Hold™ thermal transfer label kits are designed for use with glass and plastic Tubes and Vials exposed to temperatures as low as –196 °C (–320 °F).
All kits are supplied with one roll, one ribbon, and one cleaning wipe per kit.
Labels are designed to adhere to frozen surfaces including glass and plastics (wipe off frost before applying). Wrap-around design self-adheres to tubes ranging in size from 0.5 to 50 ml. Withstands temperature ranges from liquid nitrogen exposure through autoclaving. Resistant to solvents such as DMSO, methanol, and isopropyl alcohol. Labels are kitted withMore Product Information