Axygen® Automation Tips for Molecular Devices FLIPR Tetra, Corning
Supplier: Corning
FT-384-C-R FT-384-BK-R
CA10770-590CS 9438.19 CAD
CA10770-590 CA10770-592
Axygen® Automation Tips for Molecular Devices FLIPR Tetra, Corning
Pipette Tips
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Designed for use in molecular devices FLIPR tetra automation systems.
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Certified RNase-/DNase-free and non pyrogenic
  • Black and clear tips in 384-well format