Sodium hydrogen carbonate USP, FCC, Ph. Eur.
Supplier: Macco Organiques S.R.O
Synonyms:
Sodium bicarbonate
51106000455
89492-716EA
1960.37
CAD
89492-716
Sodium hydrogen carbonate USP, FCC, Ph. Eur.
Sodium hydrogen carbonate
For excipient use only.
Certifications: Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.
Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.
Formula:
NaHCO₃ MW: 84.01 g/mol |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...