Water, PCR certified DNase/RNase/Endotoxin tested, sterile

Supplier: TEKNOVA CA

Synonyms: Hydrogen oxide

W3450 W3460 W3440 W3440 W3450 W3460
CA101447-434EA 207.38 CAD
CA101447-434 101447-434 CA101447-436 101447-436 CA101447-438 101447-438
Water, PCR certified DNase/RNase/Endotoxin tested, sterile
Water

DNase, RNase and Protease tested Teknova Cell Culture Grade Water.


  • Nuclease and Endotoxin Free
  • Sterile, <0.05EU/mL. UV treated and 0.1 µm sterile filtered
  • Meets standards for ASTM Type I

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

DNase tested
RNase tested
Protease tested
PCR tested
Endotoxin tested

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR