Carts, Element Series, TRIPPNT
Supplier: TrippNT
51610 51498 51504 51609
10754-656EA 4255.56 CAD
10754-656 10754-658 10018-872 10018-874 10020-952 10020-954 10020-956 10020-958
Carts, Element Series, TRIPPNT
Trolleys
TrippNT's Element Cart Series is constructed of 2 skins of corrosion resistant aluminum with a high density polyethylene core
  • Fully customizable corrosion resistant carts using built in tracks
  • Superior maneuverability
  • Constructed with polyethylene and aluminum materials
  • Rugged, flexible, movable, workspace and storage
  • Delivered fully assembled, no tools required
  • Made in the USA
  • 100% Total Replacement Lifetime Guarantee
Order a base model and then add whatever accessories fit your work situation. All TrippNT Carts have a 100% Total Replacement Lifetime Guarantee. White color. Polyethylene and Aluminum material.
Each cart is fully customizable using built in tracks and has superior maneuverability and 4-inch nylon rubber non-marking casters.
Accessories information: Accessories can be added to any cart in the series. Add IV poles, sharps containers, trash cans, gas tanks and cardiac boards. Rugged, flexible, movable, workspace and storage. Delivered fully assembled, no tools required. Accessories can also be ordered separately to add to previously purchased carts. Made in the USA. 100% total replacement lifetime guarantee.