Hydrocarbon Soluble Siliconizing Fluid
Supplier: Thermo Scientific
TS-42801 TS-42800
10066-326EA 434.88 CAD
10066-326 10066-328
Hydrocarbon Soluble Siliconizing Fluid
Lubricants
Ideal for siliconizing glass surfaces.
  • Short chain, clear polymeric silicone fluid consists primarily of dichlorooctamethyltetrasiloxane and is soluble in organic solvents
  • Excellent for modifying metals, certain plastics, ceramics and fiber optics
When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tiightly bonded film over the entire surface.
Hydrocarbon soluble siliconizing fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid. The fluid is acidic only during application; after application, the surface is neutral.
Standard components: 1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxan
Caution: For research use only. Not for use in diagnostic procedures.