Mini-PROTEAN® TGX™ Precast Gels, Protein Gels, Bio-Rad
Supplier: Bio-Rad
4561086 4561094 4561044 4561026 4569036 4561036 4561084
101646-012EA 241.74 CAD
101646-012 101646-020 101645-996 101645-976 101646-036 101645-988 101646-008
Mini-PROTEAN® TGX™ Precast Gels, Protein Gels, Bio-Rad
Electrophoresis Gels Precast Gels
Mini-PROTEAN® TGX™ Precast Gels have been engineered for a long shelf life
  • 12 month shelf life
  • Electrophoresis run times as short as 15 min
  • Use same running and sample buffers as Laemmli systems
  • Numbered, marked well outlines on the cassette ease sample loading and identification
  • Bottom-open cassette design allows easy gel handling
  • Protein migration patterns are similar to Laemmli gels
  • Opening lever is 100% aluminum and recyclable
All Mini-PROTEAN® Gels are compatible with Mini-PROTEAN® Tetra (1–4 gels) and Mini-PROTEAN® Dodeca™ (1–12 gels) Cells. These gels can also be used in the discontinued Mini-PROTEAN® 3 Cell.
TGX™ (Tris-glycine eXtended) Gels are an innovative PAGE system designed to provide Laemmli-like separation patterns using standard Tris-glycine running buffer.