HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
Supplier: Air-Tite
5300.X00D0 5200.000V0 5050.000VZ 5200.X00V0 5050.X00V0 8300005762 5100.000V0 5100.X00V0 8300018745 8300014579 5300.000D0
89215-236CS 887.86 CAD
89215-236 89215-222 89215-226 89215-232 89215-238 89215-240 89215-234 89215-218 89215-230 10148-330 89215-217 89215-225
HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
Syringes General-Purpose Syringes
HENKE-JECT® syringes are a high quality disposable rubber-piston syringe offering substantial savings.
  • ETO gas sterilized
  • Latex and pyrogen free
A lubricated synthetic rubber piston allows for a smooth draw while a positive stop helps prevent accidental pullout. Bold graduations are easily read and resist wipe off.
Certifications: Meets ISO standards, CE registered.
Packaging: All HENKE-JECT® syringes are packaged within individual sterile strip packs.