11 mm Wide Opening Glass Crimp/Snap Top Vials, Thermo Scientific
Supplier: Thermo Scientific
C4011-88W C4011-87 C4011-87W C4011-73 C4011-92 C4011-88 C4011-5 C4011-6W
66021-192EA 331.22 CAD
66021-192 66030-126 66030-128 66030-162 66030-168 66030-170 66030-174 66030-184 66030-452 66064-194 66064-208 66064-210 66064-212 66064-216 66064-218 66064-222 66064-228 66064-230 66064-246 66064-248 66064-250 66064-252 66064-254 66064-916 66064-918 66064-920 66064-924 66064-926 66064-958 66065-260 69400-092 97054-222 66011-241 66030-130 66030-160 66030-550 66030-554 66030-556 66064-256 66064-260 89235-904 89235-950 89235-952 89235-960 89235-962 76202-158
11 mm Wide Opening Glass Crimp/Snap Top Vials, Thermo Scientific
Vials
Use Thermo Scientific 11 mm Crimp/Snap Top Vials with snap caps or aluminum crimp seal closures. These high-quality 11 mm glass vials are 2 ml, 12×32 mm and fit most brands of autosamplers. Made of superior quality 33 expansion borosilicate clear Type 1, Class A or 51A Type 1, Class B amber glass, they meet all requirements of US, EU, and JPN Pharmacopia.
Easy-to-fill wide-neck opening vials require 6 mm diameter micro-inserts.
Save time during sample preparation and reduce the risk of contamination using Thermo Scientific 11 mm Crimp Top Clear Vial Convenience Kits (Unassembled). Each kit includes 100 clear glass vials and 100 crimp top caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures, while keeping matching supplies together. Clear trays make it easy to keep track of available supplies without opening containers.
Identify sample and volume with graduated, write-on patch provided on certain vials. Get maximum sample extraction without the need for separate inserts by using Microsampling and High Recovery Vials. Obtain optimal recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional). Polyspring inserts are self-aligning and provide a cushion against needle contact. Precision point inserts minimize residual sample loss. Pulled point inserts are an economical choice for noncritical applications. Compatible with most HPLC and GC autosamplers.