Leica-type Specimen Chucks, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
SPSD22RD SPSD22BK SPSD22GR SPSD25RD SPSD25BK SPSD25BL SPSD25YL SPSD25WT SPSD25PK SPSD25GN SPSD25OR SPSD30RD SPSD30BK SPSD30BL SPSD30YL SPSD30WT SPSD30PK SPSD30GN SPSD40YL SPSD40WT SPSD22PK SPSD30OR SPSD40RD SPSD40BK SPSD40BL SPSD40PK SPSD40GN SPSD55RD SPSD55BK SPSD55BL SPSD55YL SPSD55WT SPSD55PK SPSD55GN SPSD22BL SPSD22YL SPSD22WT
10756-162EA 111.93 CAD
10756-162 10756-164 10756-166 10756-168 10756-170 10756-172 10756-174 10756-176 10756-178 10756-180 10756-190 10756-192 10756-194 10756-196 10756-198 10756-200 10756-202 10756-204 10756-206 10756-208 10756-210 10756-212 10756-214 10756-216 10756-218 10756-278 10756-280 10756-282 10756-284 10756-286 10756-288 10756-290 10756-292 10756-294 10756-304 10756-306 10756-308
Leica-type Specimen Chucks, Springside Scientific
Specimen Mounting Supplies
Suitable for Leica (except for new model CM1950), TBS, Tanner, older Avantik and other common cryostats
  • Orientation mark
  • Proper tolerance and soft aluminum, won't slip in chuck
  • Helps keep track of similar research specimens
Order Now
SPECIFICATIONS
Environmentally Preferable