Anti-CPN10 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1790
10209-964EA 1442.03 CAD
10209-964
Anti-CPN10 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for 10 kDa heat shock protein, mitochondrial(HSPE1) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: CPN10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR