Anti-AQP5 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1230
10208-008EA 1440.53 CAD
10208-008
Anti-AQP5 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Aquaporin-5(AQP5) detection. Tested with WB, IHC-P in Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: AQP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR