Anti-Nebulin Mouse Monoclonal Antibody [clone: Neb-20]

Supplier: Boster Biological Technology
MA1069
10206-066EA 1442.03 CAD
10206-066
Anti-Nebulin Mouse Monoclonal Antibody [clone: Neb-20]
Antibodies
Mouse IgG monoclonal antibody for Nebulin, nebulin (NEB) detection. Tested with WB in Human;mouse;rat;frog;chicken. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Nebulin
Clonality: monoclonal
Clone: Neb-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR