Water, Cell Culture Grade low in endotoxins

Supplier: VWR

Synonyms: Hydrogen oxide

Filtered through a 0.2 µm filter.

02-0200-0500 02-0200-1000
VWRL0200-0500CS 276.97 CAD
VWRL0200-0500 VWRL0200-1000
Water, Cell Culture Grade low in endotoxins
Water

VWR® Cell culture grade water is extremely low in endotoxin, free of DEPC (Diethyl pyrocarbonate) and other chemical additives. The product is 0.2 um filtered. It is suitable in cell culture applications.


  • Low endotoxin
  • Free of DEPC


Applications:
Media/Buffer/Reagent preparation for cell culture
Upstream Process Development Only/Reagent preparation
Downstream Process Development Only/Reagent preparation


Caution: For research and further manufacturing use. Not for Therapeutic Applications.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Conductivity before packaging ≤ 1.3 uS/cm
Endotoxin (EQC) ≤ 0.25 EU/ml
Microorganism Growth (EQC) No Growth
pH (EQC) Passes test
Total Organic Carbon < 500 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR