Uniguard™ Direct-Connection Guard Cartridge Holders, Thermo Scientific
Supplier: Thermo Scientific
850-00 852-00 851-00
10046-748EA 457.7 CAD
10046-748 10046-746 10046-744
Uniguard™ Direct-Connection Guard Cartridge Holders, Thermo Scientific
Guard Column Holders
Eliminate the requirement for extra fittings using Thermo Scientific Uniguard Direct-Connection Guard Cartridge Holders
With PEEK 0.0625" male outlet that fits many columns; 0.0625" female inlet tip can be used with various standard fittings.
They are reusable, stainless steel guard cartridge holders that attach directly to the analytical column inlet.