VWR® Roller Bottles, 1000 and 2000 ml, Sterilized, Non-Pyrogenic, Standard Line

Supplier: VWR International
10769-794 10769-796 10769-800 10769-802
10769-794CS 402.24 CAD
10769-794 10769-796 10769-800 10769-802
VWR® Roller Bottles, 1000 and 2000 ml, Sterilized, Non-Pyrogenic, Standard Line
Bottles Roller Bottles
Roller Bottles are optimal vessels for a large yield of cell growth and multiplication, to be used in both research and manufacturing applications.

  • Available in two sizes: 1000 and 2000 ml
  • Available surface treated
  • Caps feature large knurls for easy grip
  • Bottle surface is smooth, free of striations
  • Sterilized by electron beam irradiation
  • Non-pyrogenic, not autoclavable
  • Temperature range: –20 to 50 °C (–4 to 122 °F)

Two different cap styles can be used in both open and closed systems. Suitable for all common instruments and automation. Every caseMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR