Calibrated 1000 Series GASTIGHT® Syringes, Hamilton
Supplier: Hamilton
CAL80200 CAL80900 CAL81000 CAL81100 CAL81217 CAL81317 CAL81417 CAL81517
89232-390EA 472.52 CAD
89232-390 89232-392 89232-394 89232-396 89232-398 89232-400 89232-402 89232-404
Calibrated 1000 Series GASTIGHT® Syringes, Hamilton
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. The Calibrated 1000 series is a mid volume Gastight® syringe. A Certificate of Calibration is provided with the product and the procedure is performed with an unbroken chain of calibrations traceable to N.I.S.T. Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze. This series of syringes is ideal for dispensing volumes from 100 μL up to 100 mL. A variety of different terminations and needle options are available.
1000 Series Syringe Terminations–Luer Tip Cemented Needle (LTN)–For mid volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
Removable Needle (RN)–The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Luer Tip (LT)–The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
PTFE Luer Lock (TLL)–This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
Certifications: N.I.S.T.
Ordering information: For a needle point or gauge that is available but not listed here, please contact your Avantor representative to place a special order by phone.