Perform, Nitrile Exam Gloves, Supermax Healthcare Canada
Supplier: SUPERMAX HEALTHCARE CANADA CA
92395 92396
CA10500-782CS 615.15 CAD
CA10500-782 CA10500-784 CA10500-790
Perform, Nitrile Exam Gloves, Supermax Healthcare Canada
Gloves
Optimal stretchability, just like latex.
  • Beaded cuff snugs wrist, eliminates roll-up, minimizes entry of liquids
  • Long-lasting comfort, durability and decreased hand fatigue
  • Textured fingertips for precision gripping
  • Hypoallergenic
  • Protect hands from biohazards and a broad range of chemicals
Certifications: ISO 9001: 2000 certified by SGS (UK) and SIRIM (Malaysia); ISO 13485: 2003 certified by SGS (UK); CMDCAS ISO 13485 Certification.
Ordering information: Packaged 200 gloves per dispenser box.