Manual GC Syringes for GC Autosampler, Thermo Scientific
Supplier: Thermo Scientific
365B0511 365B0531
10055-188EA 292.9 CAD
10055-188 10055-186
Manual GC Syringes for GC Autosampler, Thermo Scientific
Syringes Syringes for Chromatography
Achieve accurate, precise, and reproducible manual injections easily using Thermo Scientific™ Manual GC Syringes
The robust, field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes. Easy-to-read markings ensure reproducibility.
Accessories information: Thermo Scientific™ Repair Kits for Thermo Scientific Plunger-in-Needle Syringes extend the life and performance and reduce cost per use of syringes.