Stratos® Lens, MCR Safety
Supplier: MCR Safety
SS110 SS112 SS010 SS120
47732-278PK 116.11 CAD
47732-278 47732-280 80082-004 80082-012
Stratos® Lens, MCR Safety
Safety Glasses
The Stratos® features a one piece polycarbonate lens and a stylish wrap-around design that offers the user a wide area of protection.
  • Integral side shields offer protection from particles entering the eyewear from the sides
  • Molded-in nose bridge comfortably fits a wide variety of faces
  • Available in four frame and two lens tint options
The Stratos® provides an exceptional wide angle of unrestricted vision allowing you to safely assess your environment. The Stratos® features a stylish wrap-around design and one piece polycarbonate lens with integral sideshields that filters out 99.9 percent of UV radiation. The lightweight frame provides added comfort and balance.