Lens Cleaning Station, MCR Safety
Supplier: MCR Safety
LCS1
80081-970PK 287.12 CAD
80081-970
Lens Cleaning Station, MCR Safety
Cleaning Agents Lens Cleaning Solutions
This cleaning station contains 2 boxes of 300 tissues and one 16-ounce bottle of anti-fog, anti-static cleaning spray. It assist in the care and maintenance of your safety eyewear.
  • For plastic, PC and glass lenses
  • Anti-fog, anti-static formula
  • Contains no silicone
Ideal for glasses, goggles, face shields and computer screens.