Yukon®Visitor Spec, Dielectric, MCR Safety
Supplier: MCR Safety
9800B 9800D 9810 98120 98150 9800XL 9800XLB 9800XLD 9810XL
66025-228PK 148.84 CAD
66025-228 66025-230 66025-238 66025-240 66025-262 66025-266 66025-270 66025-272 66025-274 66025-276 66025-278 66025-280
Yukon®Visitor Spec, Dielectric, MCR Safety
Safety Glasses
The Yukon® features a contemporary, dielectric wrap-around style.
  • Excellent option for visitor glasses
  • Dielectric, no metal parts
  • Rugged polycarbonate lens filters 99.9% of harmful UV rays
  • Single lens design with integral side shields for greater orbital seal
  • Spatula temples
It provides increased angular coverage and improved peripheral vision. Molded-in brow guard and vented sideshields give additional protection for welding. Available in both hard-coated for scratch resistance, or uncoated for extra cost savings.
Certifications: Meets or exceeds ANSI Z87+ and CSA Z94.3.