Micro-Splitter Valve, Upchurch Scientific®
Supplier: IDEX Health & Science
P-450
53500-630EA 265.96 CAD
53500-630
Micro-Splitter Valve, Upchurch Scientific®
Valves Micro Valves
Designed to accurately split and control a low-flow stream of a single incoming supply.
  • For interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow ratesFor interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow ratesFor interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow rates