RheBuild® Kits, Rheodyne®, IDEX Health & Science
Supplier: IDEX Health & Science
7960-999 7960-999 7739-999
97058-342EA 326.6 CAD
97058-354 97058-342 97058-356
RheBuild® Kits, Rheodyne®, IDEX Health & Science
Chromatography Supplies Chromatography Spare Parts
RheBuild® kits containing genuine Rheodyne® parts are available for most Rheodyne® products
RheBuild® kits eliminate the need to order individual parts.
Ordering information: Each RheBuild® kit includes all the parts, tools and instructions to maintain the precision performance of Rheodyne® valve platforms.