Rheodyne® MXP High Pressure Valves, MX Series II™, IDEX Health & Science
Supplier: Rheodyne
MXP7900-000 MXP7960-000 MXP9900-000
97058-376EA 6634.18 CAD
97058-376 97058-380 97058-382 97058-388 97058-390
Rheodyne® MXP High Pressure Valves, MX Series II™, IDEX Health & Science
Valves Switching Valves
Chemically compatible for use with most mobile phase compositions.
  • Can withstand up to 6000 psi (414 bar)
  • Available with analytical and nano-scale flow paths
  • Multiple control options
  • Stand alone capability
  • Included software