Rheodyne® MXX Low Pressure Valves, MX Series II™, IDEX Health & Science
Supplier: Rheodyne
MXX777-605 MXX778-605
97058-396EA 4194.6 CAD
97058-396 97058-400 97058-402 97058-404 97058-406
Rheodyne® MXX Low Pressure Valves, MX Series II™, IDEX Health & Science
Valves Switching Valves
Combine MX Series II™ modular valves with an instrument currently in use to support complex fluid switching and sample injection needs.
  • Can withstand up to 125 psi (9 bar)
  • All models accept 1/16" or 1/8" tubing (proper ferrules required)
  • Multiple control options
  • Stand alone capability
  • Included software