WHEATON® MicroLiter 10-425 Screw-Thread Vials, Certified Clean, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
10-1000 10-1200 10-1200-300 10-2200-101 10-2200-S 10-1000-S
97046-953PK 241.01 CAD
97046-953 97046-956 97046-971 97046-972 97046-973 97046-974 97046-991 97046-993 97046-995 97046-955
WHEATON® MicroLiter 10-425 Screw-Thread Vials, Certified Clean, DWK Life Sciences
Vials
All vials may appear to be created equal, but quality and performance make the difference when analyzing vital samples.
  • Identical and dependable
MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials. Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present.
MicroLiter vials are identical, clean, dependable vials, certified to ensureMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable