Dominator Jacket, Attached Hood, MCR Safety
Supplier: MCR Safety
388JHS 388JHM 388JHL 388JHXL 388JHX2 388JHX3 388JHX4 388JHX5
89066-086EA 135.66 CAD
89066-086 89066-088 89066-090 89066-092 89066-094 89066-096 89066-098 89066-100
Dominator Jacket, Attached Hood, MCR Safety
Workwear
The Dominator Series is made tough and roomy for reliable use in dirty jobs
It is easily identified by its kelly green color and has additional features that help protect you against splash, grit, and grime. The 388JH is a jacket with attached drawstring hood. It also features a zipper front with oversized storm flap and take up snaps at the wrists. This jacket has a plain back, no pockets, and no vents to protect from splash. This material is treated to be limited flammability. Dominate your environment with the Dominator.
Order Now
SPECIFICATIONS
Environmentally Preferable
Garment application General Purpose
Risk area
Gender Unisex
Sleeve or leg length Long
Closure type Zip closure
Fabric PVC/nylon
Thickness 0.42 mm