8-425 Glass Screw-Thread Vials, Thermo Scientific
Supplier: Thermo Scientific
C4013-2 C4013-015A C4013-15 C4013-58AW C4013-1 C4013-1W C4013-2W
66001-239PK 115.81 CAD
66001-239 66030-422 66030-424 66030-426 66030-432 66030-434 66030-436 66030-438 66030-442 66030-450 66064-276 66064-284 66064-288 66064-302 66064-304 66065-200 66065-204 66065-208 66065-216 66065-312 69400-088 69400-094 66001-236 66065-120 66065-122
8-425 Glass Screw-Thread Vials, Thermo Scientific
Vials
Compatible with a broad variety of autosamplers, these Thermo Scientific 8 mm Glass Screw Thread Vials have a 8-425 thread finish.
  • Polyspring inserts are self-aligning and provide a cushion against needle contact
  • Precision point insert minimizes residual sample loss
  • Pulled point inserts are an economical choice for noncritical applications
Save time during sample preparation and reduce the risk of contamination with Thermo Scientific 8 mm Vial Kits, assembled or unassembled, which include 100 glass vials preassembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers or in economical polybags. Available as silanized (deactivated) for optimal recovery of critical polar, labile or OH-interacting compounds.