Hei-VAC Valve Control, Hei-VAC Valve Tec Valve Regulated Vacuum Pumps
Supplier: Heidolph
Hei-VAC Valve Control, Hei-VAC Valve Tec
36304780 036302830 036304781 36304900 036304780CA
CA10145-620EA 5432.8 CAD
89496-052 14224-096 CA10145-620 89428-204 10811-482
Hei-VAC Valve Control, Hei-VAC Valve Tec Valve Regulated Vacuum Pumps
Pumps Vacuum Rotary Evaporator Pumps
These valve-regulated vacuum pumps can be used with all Hei-VAP rotary evaporator models.
  • Vacuum can be controlled via valve operated or manual vacuum controllers
  • Depending on your application you can switch on and off the vacuum pump via switchbox (only for Hei-VAC Valve Control)
Delivery information: Condenser is not in scope of delivery.