13-425 Screw-Thread Vials
Supplier: Thermo Scientific
C4015-17 C4015-17W C4015-17A C4015-17AW C4015-21W C4015-21AW C4015-017 C4015-017A C4015-88 C4015-88AW C4015-21 C4015-14 C4015-9
66030-478PK 272.92 CAD
66030-478 66030-480 66030-482 66030-484 66030-488 66030-492 66030-494 66030-496 66030-498 66030-508 66030-510 66030-516 66033-022 66064-328 66064-330 66064-332 66064-334 66065-138 66065-142 66065-222 66065-674 69400-078 69400-080 66030-380 66030-382 66030-384 66030-386
13-425 Screw-Thread Vials
Vials
These 4 ml, 15×45 mm vials feature a 13-425 thread finish.
Thermo Scientific National unassembled 4 ml screw thread vial convenience kits include 100 vials and 100 caps with pre-assembled septa.
These vials are made of borosilicate glass material.
All items remain handy and dust-free. Caps with pre-inserted septa are not attached to vials; packaged in polybags.