SP Bel-Art Magnetic Bead Separation Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19900-0000 F19900-0001 F19900-0002 F19900-0003 F19900-0004
10770-234EA 752.13 CAD
10770-234 10770-236 10770-238 10770-240 10770-242
SP Bel-Art Magnetic Bead Separation Racks, Bel-Art Products, a part of SP
Racks Magnetic Separation Racks
Designed for use in all magnetic separation experiments, these powerful racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin, or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays.
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves
  • Simply turn tube for efficient bead mixing
  • Racks fit microplates following ANSI standards
  • Clear acrylic allows unobstructed view of tube contents
Magnetic Bead Separation Racks offer exceptional quality and value compared toMore Product Information