DigiTUBE® Digestion Tubes
Supplier: Revvity Health Sciences
N9308008 N9308066 N9308016 N9308037
97050-124EA 1582.06 CAD
97050-124 97050-066 97050-082 97050-182
DigiTUBE® Digestion Tubes
Tubes Digestion Tubes
Disposable sample digestion tubes with Class A graduations.
  • Easy-to-read graduations
  • Allows normalization directly in the tube
  • Easy one-handed closure of leak-free screw cap
All-in-one tubes eliminate the need for five separate vessels such as Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.