Cleanroom Barrier Tapes, Micronova

Supplier: Micronova
BT200-3YE BT100-3RE
89236-626CS 1016.95 CAD
89236-626 89236-628
Cleanroom Barrier Tapes, Micronova
Tapes
Tapes made of high visibility, non-adhesive polyethylene film. Ideal for blocking restricted areas and recommending caution in controlled environments.

  • Text is in English and Spanish (2" hgh lettering)
  • Yellow or red film with bold black print
  • Wound on a 76 mm (3") plastic core

These tapes help prevent accidents by increasing visibility of hazards and can be tied, stapled or nailed in place.

Length: 304.8 m (1000 ft.)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR