Stainless Steel Wire Brushes and Covers, Micronova
Supplier: Micronova
BRWM-125 BRWM-22 BRWL-349 BRWM-22512S BRWM-341512 BRWM-345 BRWL-149 BRWL-14349 BRWM-16BR BRWM-12572 BRWM-1572 BRSC6-156
10126-338CS 1013.87 CAD
10126-338 10126-348 10753-442 76415-986 10126-332 10126-330 10126-340 10126-336 10126-346 10126-368 10126-334 10126-344
Stainless Steel Wire Brushes and Covers, Micronova
Brushes
The Stainless Steel Wire Brushes with nylon bristles are excellent for cleaning valves, tubing, cylinders, narrow chambers, and other hard to clean areas.
  • Nylon bristle brushes with a stainless steel wire core
  • Ideal for valves, tubing, cylinders, and narrow chambers
  • Flexible to bend through fittings but stiff enough to remove residues
  • Available in a range of diameters, widths, lengths, and bristle stiffness
  • Option for looped or plastic end
  • Made from FDA compliant materials
The durable brushes are flexible enough bend through fittings but stiff enough for removing residues. The tapered versions add a level of cleaning versatility.
Certifications: FDA Approved Materials.