Autoclave Indicator Tape, Lead Free, Micronova
Supplier: Micronova
AUTOPC-1 AUTOPC-25 AUTOPC-3 AUTOPC-34 AUTOPC-15 AUTOPC-2
10147-800CS 1858.79 CAD
10147-800 10152-758 10152-760 10153-150 10153-152 10153-154
Autoclave Indicator Tape, Lead Free, Micronova
Tapes
Method of positively confirming complete sterilization cycle in both gravity discharge and vacuum autoclaves.
  • Lead and latex-free
  • Wound on plastic core
  • Easy to handle tape
The green indicator lines change to black when proper levels of steam pressure and temperature have been achieved. The uniquely printed lines cannot be rubbed off and will not contaminate instruments. After use, the tape removes quickly and completely, leaving minimal residue or staining. Tape adheres on contact and stays in place through live steam pressure.
Certifications: RoHS compliant.
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health