KIMAX® Specific Gravity Bottles, Pycnometer, DWK Life Sciences
Supplier: DWK Life Sciences
15123N-25 15123N-50 15123N-10
10146-726EA 611.83 CAD
10146-646 10146-648 10146-726
KIMAX® Specific Gravity Bottles, Pycnometer, DWK Life Sciences
Specific Gravity Bottles
Pycnometer bottles are provided with non-mercury, red-spirit filled thermometers for facilities that are designated mercury-free.
Bottles are designed with a special, medium length [ST] 10/18 joint and are retested for accuracy within ±0.2 °C. Cap for side tube is medium length [ST] 5/12 and has a small vent near the top. Body is conical for maximum stability and capacity is within 10 percent of nominal shown on the flask. Each flask body is numbered for ready identification. The 50 mL size is indicated in the determination of the specific gravity of pigments (ASTM D153).