Sodium dodecyl sulfate (SDS) 99% for molecular biology

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

J67606-22 J67606-30 J67606-36
CAAAJ67606-22EA 121.5 CAD
CAAAJ67606-22 CAAAJ67606-30 CAAAJ67606-36
Sodium dodecyl sulfate (SDS) 99% for molecular biology
Sodium dodecyl sulphate (SDS)

Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade. Solid.


Purity: 99% CAS No.: 151-21-3 Molecular Formula: C12H25NaO4S Formula Weight: 288.38 Physical Form: Solid Appearance: White Melting Point: ca 206° Solubility: Soluble in water Beilstein Reference: 3599286 UN No.: 2926 MDL No.: MFCD00036175

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: ∼ 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,8636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR